Learn More
Brady™ Polyester Labels for Thermal Transfer Printers
GREENER_CHOICE
For barcoding and identification purposes
Brand:  Brady™ 116372
 View more versions of this product
Product Code. 12678293
  • 155.00 € / 1 roll
Explore more special offers
Description
Description
  • Available with gloss or matte finish
  • Available in different widths and roll lengths
Specifications
Specifications
RoHS 2002/95/EC
Acrylic, Permanent
Matte
Polyester
BBP™11 Label Printer
1500/Pk.
1.9 x 5.59 cm (W x H)
0.048 mm
0.5 in.
12.70 mm
5.59 cm
1 in.
Thermal Transfer Printable Labels
White, Translucent
Label
Laboratory identification such as vials, centrifuge tubes, test tubes, straws, and slides
1
Rectangle
-196 °C to 110 °C
15.88 mm
1.9 cm
2.2 in.
19.05 mm
2.54 cm