Learn More
Brady™ Polyester Labels for Thermal Transfer Printers
For barcoding and identification purposes
Brand:  Brady™ 116410
 View more versions of this product
Product Code. 30041400
  • 656.00 € / Pack of 2500
Explore more special offers
Description
Description
  • Available with gloss or matte finish
  • Available in different widths and roll lengths
Specifications
Specifications
RoHS 2002/95/EC
Acrylic, Permanent
Matte
Polyester
BBP™11 Label Printer
6.35 W x 6.35 cm H
6.35 cm
6.35 cm
THT-142-481-2.5-SC
White
Label
Laboratory identification such as vials, centrifuge tubes, test tubes, straws, and slides
2500/Pk.
2.5 in.
2.5 in.