Learn More
Bollé Safety Polyester Neck Cord
Keep track of your Bolle Protection™ safety glasses with the Polyester Neck Cord, a helpful accessory.
Brand: Bollé Safety CORDC
15.95 EUR valid until 2025-06-08
Use promo code "24131" to get your promotional price.
Description
- Polyester

Specifications
Blue, green | |
Bollé Safety Spectacles | |
CORDC |
Neck Cord | |
560 mm | |
Polyester |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.