Learn More
Macherey-Nagel™ Polyester Sheets Polygram SIL G, CEL 300 TLC Micro-Set F1
Brand:  Macherey-Nagel™ 814200
Product Code. 12430107
  • 272.00 € / Each
Explore more special offers
Specifications
Polyester Sheets Polygram SIL G, CEL 300
TLC Micro-Set F1
4 x 8 cm
1 Ea.
1 manual; 50 glass capillaries 1 μL; 50 polyester sheets each of Polygram SIL G/UV254 and CEL 300, 4 x 8 cm; 100 mL each of n-butanol, ninhydrin spray reagent (in ethanol), acetone, 25 % ammonia solution, rubeanic acid spray reagent (in ethanol)*; 50 mL each of 50 % acetic acid, 18 % hydrochloric acid*; 8 mL each of the amino acid test mixture (see above), arginine reference solution and tryptophan reference solution (each aq. soln.); 8 mL each of the heavy metal cation test mixture (see above), Mn(II) reference solution and Cu(II) reference solution (each aq. soln.)
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • TLC microset F1
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • H400-Very toxic to aquatic life.
  • H318-Causes serious eye damage.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : acetic acid, acetone, ammonia solution,manganese(II) sulphate monohydrate, copper(II) sulphate pent