Learn More
Brady™ Polyester Thermal-Transfer Printable Labels
Polyester Thermal-Transfer Printable Labels are made of permanent polyester (B-423), which is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.
Brand:  Brady™ 217092
 View more versions of this product
Product Code. 10561515
  • 631.00 € / 1 roll
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Glossy white finish with permanent acrylic is thermal transfer printable
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
  • Meets requirements of a halogen-free material per DIN VDE 0472 part 815
  • Service temperature of -94 to 230° F (-70 to 110° C)
Specifications
Specifications
2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Thermal Transfer Printable Labels
White
Self-laminating
Thermal Transfer
Laboratory identification such as vials, centrifuge tubes, test tubes, straws, and slides
1
Rectangle
-196°C to +110°C
25.40 x 48.26 mm
48.26 mm
25.4 mm
BPT-636-461-AW
Acrylic, Permanent
Matte
Polyester
Thermal Transfer Printer Label
BBP11, BP1244, BP1344, BP4000, BP4320
2000/Pk.
18.26W x 25.46 mmH
0.012 mm
18.26 mm
38.1 mm