Learn More
Brady™ Polyester Thermal-Transfer Printable Labels
Polyester Thermal-Transfer Printable Labels are made of permanent polyester (B-423), which is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.
Brand:  Brady™ 031337
 View more versions of this product
Product Code. 13009312
  • 265.00 € / 1 pack
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Glossy white finish with permanent acrylic is thermal transfer printable
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
  • Meets requirements of a halogen-free material per DIN VDE 0472 part 815
  • Service temperature of -94 to 230° F (-70 to 110° C)
Specifications
Specifications
RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC, UL Recognized, CSA Accepted
Thermal Transfer Printable Labels
Powder
Gloss
Polyester
Thermal Transfer Printer Label
BBP11, BP1244, BP1344, BP2461, BP3481, BP6441, MVP200, MVP300, 300XPlusII, 360XPlusII, 600XPlusII, PR200Plus, PR300Plus, PR600Plus, IP300, IP600
10,000/Pk.
25.4W x 12.7mmH
0.025 mm
12.70 x 25.40 mm
12.7 mm
THT-59-483-10
Rubber, Permanent
White
IP™ Enabled
Thermal Transfer
Asset identification, rating and nameplate labels
1
Rectangle
-40°C to +120°C
12.24 mm
25.4 mm