missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyester Wiper
Polyester Wiper, Length: 230 mm, White, Packaging Quantity: 150Pack, Width: 230mm
Brand: Texwipe 33677-00
This item is not returnable.
View return policy
Description
Polyester Wiper, Length: 230 mm, White, Packaging Quantity: 150Pack, Width: 230mm
Specifications
Wiper | |
230 mm | |
White |
230 mm | |
Polyester | |
150 Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction