Learn More
Masterflex™ Polyethylene Barbed Y-Connectors
Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing
Brand:  Masterflex™ 30726-07
 View more versions of this product
Product Code. 12848100
  • 62.20 € / Pack of 10
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Barbed Y-Connectors
  • Polyethylene
  • Available in pack of ten
Specifications
Specifications
Barbed Y-Connector
Not Autoclavable
1/2 in.
1/2 in.
Ethylene Oxide
Barb Connections
HDPE
1/2 in.
1/2 in.
Translucent