missing translation for 'onlineSavingsMsg'
Learn More

Alpha Packaging™ Polyethylene Biohazard Bags

Product Code. p-8007820
Click to view available options
Width (Metric):
140 mm
150 mm
Unit Size:
Pack of 100

Product Code. 12390529

Brand: Alpha Packaging™ 12390529

Only null left
Add to basket
Edge
Add to basket

Use either plain or printed Alpha Packaging™ Polyethylene Biohazard Bags for specimen collection and transport. They are printed with a biohazard warning.

  • Plain or printed with a biohazard warning
  • Opening along width. 50μm thickness
  • Resealable section for the specimen

Specifications

Material Polyethylene
Thickness (Metric) 50 μm
Width (Metric) 150 mm
Product Type Biohazard Bag
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.