missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Polyethylene Bung

Manufactured with polyethylene for strength, durability and flexibility. Buerkle™ Polyethylene Bungs are available in a wide range of sizes.

0.75 € - 3.85 €

Specifications

Color White, Red
Material Polyethylene
Product Type Bung
For Use With (Application) For Foodstuffs
View More Specs

Products 9
Product Code Brand Standard Taper Joint Price Quantity & Availability  
Product Code Brand Standard Taper Joint Price Quantity & Availability  
11888172
View Documents
Buerkle™
9760-1926
19/26
1.20 €
Each
In Stock
to see stock.
Add to basket
 
15445574
View Documents
Buerkle™
9760-2932
29/32
1.70 €
Each
In Stock
to see stock.
Edge
Add to basket
 
11888182
View Documents
Buerkle™
9760-3435
34/35
3.20 €
Each
In Stock
to see stock.
Add to basket
 
15376438
View Documents
Buerkle™
9760-1423
14/23
1.15 €
Each
In Stock
to see stock.
Add to basket
 
11878182
View Documents
Buerkle™
9760-4045
45/40
3.85 €
Each
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
 
15337378
View Documents
Buerkle™
9760-2429
24/29
1.35 €
Each
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
 
15405574
View Documents
Buerkle™
9760-1019
10/19
1.00 €
Each
In Stock
to see stock.
Add to basket
 
15435574
View Documents
Buerkle™
9760-1221
12/21
0.95 €
Each
In Stock
to see stock.
Add to basket
 
15415574
View Documents
Buerkle™
9760-0716
7/16
0.75 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

  • Hollow stopper allows for use of tubes or funnels
  • Sealing rings provide a tight seal
  • Suitable for foodstuff
  • Octagonal shape facilitates easy opening
  • Polyethylene provides good chemical resistance
Specifications

Specifications

White, Red
Polyethylene
Bung
For Foodstuffs
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.