missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Polyethylene Elbow Connector Barbed Fitting

Create 90° bends in flexible tubing in the laboratory without kinks or the use of tools.

Brand:  Masterflex™ 30622-80

 View more versions of this product

Product Code. 11705569

  • 25.70 € / Pack of 10
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Slip flexible tubing or hose onto the barb to create a seal that typically holds pressures up to 30 psi (2 bar)
  • Available in a range of sizes
  • Manufactured using 100% virgin raw materials
  • Provided in packs of 10
Specifications

Specifications

Barbed Fitting
1/2 in. tubing I.D.
Ethylene Oxide
Polyethylene
10 Pack
Elbow
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.