Learn More
Polyethylene Glycol Antibody (RM105), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26041F
Product Code. 30027850
  • 870.00 € / 0.10mL
Explore more special offers
Description
Description
Polyethylene Glycol Monoclonal antibody specifically detects Polyethylene Glycol in All samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
Polyethylene Glycol
Monoclonal
FITC
PBS
Rabbit
Protein A purified
RUO
All species
Purified
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
RM105
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
1,2-Dihydroxyethane, 1,2-ethanediol, 107-21-1, 2-hydroxyethanol, Ethane-1,2-diol, glycol, Glycol alcohol, monoethylene glycol
KLH conjugated Polyethylene Glycol (PEG) with terminal methoxy group
0.1 mL
Primary
Store at 4C in the dark.