missing translation for 'onlineSavingsMsg'
Learn More
Polyethylene glycol dimethanesulfonate, M.W. 1,000, Thermo Scientific™ Product Code.: 15482757

Polyethylene glycol dimethanesulfonate, M.W. 1,000, Thermo Scientific™

Product Code. 15482757
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15482757

Brand: Thermo Scientific Alfa Aesar 046750.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52410-74-9
Molecular Formula C4H10O6S2
Molecular Weight (g/mol) 218.238
MDL Number MFCD03100947
InChI Key QSQFARNGNIZGAW-UHFFFAOYSA-N
Synonym ethylene dimethanesulfonate, dmse, 1,2-bis mesyloxy ethane, ethylene methanesulfonate, ethylene glycol dimesylate, ethane dimethane sulphonate, 1,2-ethanediyl dimethanesulfonate, dimethylene dimethanesulfonate, ethylene bis methanesulfonate, ethane dimethanesulphonate
PubChem CID 20796
IUPAC Name 2-methylsulfonyloxyethyl methanesulfonate
SMILES CS(=O)(=O)OCCOS(=O)(=O)C

Specifications

Color White
Physical Form Waxy Solid
Quantity 5 g
Chemical Name or Material Polyethylene glycol dimethanesulfonate, M.W. 1,000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Polyethylene glycol dimethanesulfonate, M.W. 1,000, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.