missing translation for 'onlineSavingsMsg'
Learn More
Polyethylene glycol monomethyl ether methanesulfonate, M.W. 13,000, Thermo Scientific™ Product Code.: 15477285

Polyethylene glycol monomethyl ether methanesulfonate, M.W. 13,000, Thermo Scientific™

Product Code. 15477285
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15477285

Brand: Thermo Scientific Alfa Aesar 046693.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175172-61-9
Molecular Formula C4H10O4S
Molecular Weight (g/mol) 154.18
MDL Number MFCD03100948
InChI Key BCKAHDGFNHDQST-UHFFFAOYSA-N
Synonym methanesulfonic acid 2-methoxyethyl ester, ethanol, 2-methoxy-, methanesulfonate, 2-methoxyethanol methanesulfonate, 2-methoxyethyl mesylate, 2-methoxyethylmethanesulfonate, acmc-1c990, 2-methoxyethyl methane sulfonate, methanesulfonic acid2-methoxyethyl ester, ethanol, 2-methoxy-, 1-methanesulfonate, methanesulfonic acid 2-methoxy-ethyl esterShow MoreShow Less
PubChem CID 146293
IUPAC Name 2-methoxyethyl methanesulfonate
SMILES COCCOS(=O)(=O)C

Specifications

Color White
Physical Form Granular Solid
Quantity 250 mg
Chemical Name or Material Polyethylene glycol monomethyl ether methanesulfonate, M.W. 13,000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Polyethylene glycol monomethyl ether methanesulfonate, M.W. 13,000, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.