missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Polyethylene Molded Fume Hood

This fume hood features a one-piece shell molded of polyethylene plastic with a clear acrylic sash that slides on inside tracks

Brand:  Bel-Art™ SP Scienceware™ H50000-0003

Product Code. 11787375

  • 1515.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Sash can be set in an open, closed or intermediate position; remains open slightly when in closed position to facilitate continuous air flow
  • Front opening is 88 x 36cmH (34½ x 14³/₁₆ in.)
  • Stack O.D.: 15cm (6 in.)
  • Exhaust system capable of drawing 350 cfm (free air) is recommended
  • Overall dimensions: 107 x 51 x 76cmH (42 x 20 x 30 in.)
Specifications

Specifications

Polyethylene, Acrylic
107 x 51 x 76 cm (42 x 20 x 30 in.)
14.19 in.
88 x 36 cm
20 in.
White
30 in.
76 cm
1 Ea.
51 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ SP Scienceware™ Polyethylene Molded Fume Hood > Material: Polyethylene

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.