missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Polyethylene Push-in Caps for Glass Specimen Vials
Fit securely into place to seal glass specimen vials. Fisherbrand™ Polyethylene Push-in Caps for Glass Specimen Vials are available in a variety of sizes specifically designed to work with Fisherbrand™ Push-in Cap Glass Specimen Vials.
25.35 € - 45.65 €
Product Code | Brand | Closure Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Price | Quantity & Availability | ||||||
15354789
|
Fisherbrand™
15354789 |
22 mm |
45.65 €
Pack of 300 |
In Stock
Log in to see stock. ![]() |
||||||
15364789
|
Fisherbrand™
15364789 |
8 mm |
25.35 €
Pack of 300 |
In Stock
Log in to see stock. ![]() |
||||||
15374789
|
Fisherbrand™
15374789 |
10 mm |
32.70 €
Pack of 300 |
In Stock
Log in to see stock. ![]() |
||||||
Description
- Specifically designed for use with Fisherbrand™ Push-in Cap Glass Specimen Vials
- Polyethylene construction, natural color
- Available in a variety of sizes to match vial specifications
- Push-in cap specimen vials are suitable for media, diagnostic, storage, display, and sample collection applications
Spécification
Natural | |
Polyethylene | |
Push-in Cap |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Fisherbrand™ Polyethylene Push-in Caps for Glass Specimen Vials