missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Polyethylene Round All Purpose Boxes

Box for storing, packing, taking samples, sampling and transport of pastes, granules, powders, soil samples and frozen goods (-40 °C up to briefly +95 °C). Ideal use of space, can be stacked. Complete with press-in lid

Brand:  Buerkle™ 6210-0250

3.96 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10286363

  • 4.80 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • PE, transparent
  • No crevices
  • |PE, transparent
Specifications

Specifications

250 mL
Transparent
Round
Press-In Lid
90 mm
Press-in Lid
Polyethylene
Storing, Packing, Taking Samples, Sampling and Transport of Pastes, Granules, Powders, Soil Samples and Frozen Goods (-40°C up to Briefly +95°C)
Multi-Purpose Box
78 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers