missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Polyethylene Sample Tubes

Made of lightweight and unbreakable polyethylene. Kartell™ Polyethylene Sample Tubes can be used as weighing containers.

Brand:  Kartell™ 0073700

55.87 EUR valid until 2024-12-31
Use promo code "21960" to get your promotional price.


 View more versions of this product

Product Code. 10737152

  • 72.20 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • General purpose containers with push in hinged lids
  • Flat bottom, lightweight and shatterproof
  • Wall thickness approximately 1mm
  • Suitable for foodstuff
Specifications

Specifications

5 mm
5 mL
1
50 mm
Tube
Polyethylene
Can be used as Weighing Containers
Flat Bottom
100 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.