Learn More
Masterflex™ Polyethylene Straight Connector
Designed for quick and easy assembly without tools
Brand: Masterflex™ 30621-83
This item is not returnable.
View return policy
Description
- Ideal for low-pressure applications
- Chemical resistant
- Available in a variety of sizes
- Economical barbed fittings easily connect fluid paths without the use of tools
- Just slip your flexible tubing or hose onto the barb to create a secure seal
Specifications
Straight Connector | |
Not Autoclavable | |
1 in. | |
1 in. | |
Translucent |
Barb Connection | |
HDPE | |
1 in. | |
Ethylene Oxide |