Learn More
GUERIN PLASTIQUES Polyethylene Sleeves
Use for food handling and general applications. GUERIN PLASTIQUES Polyethylene Sleeves are made of transparent, 0.1mm thick polyethylene.
Brand: GUERIN PLASTIQUES 50153
Description
- 350m long in total
- Available in a variety of widths
Specifications
1/Pk. | |
Polyethylene | |
350 m | |
Transparent |
Sleeve | |
Food Quality | |
0.1 Thick x 120Wmm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.