missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Polyethylene Wide Mouth Wash Bottle

Product Code. 12388118
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Closure Color:
Blue
Green
Natural
Red
Chemical Label:
70% Ethanol
Acetone
Custom Write-on
Ethyl Acetate
Machine Oil
Methyl Ethyl Ketone
Soap
Toluene
Water
This item is not returnable. View return policy
This item is not returnable. View return policy

Manufactured from polypropylene. Bel Art™ Polyethylene Wide Mouth Wash Bottle feature a wide mouth 53mm (2 in.) for easy refilling.

  • Pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
  • Translucent LDPE bottles; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Color-coded polypropylene closures aid in quick and easy identification
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
  • 500ml - 4/bag, 1000ml - 4/bag; assortment is 6 per bag, includes one each: Acetone, Bleach, Distilled Water, Ethanol, Isopropanol, and LYOB


Note: Use LYOB only with solvents compatible with LDPE

Specifications

Capacity (Metric) 500 mL
Color Translucent
Diameter (Metric) Outer Neck 53mm
Includes Red Polypropylene Cap
Material Polyethylene
Quantity 4Pack
Closure Color Red
Certifications/Compliance Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Mouth Wide
Product Type Wash Bottle
Chemical Label Toluene
Closure Material Polypropylene
Closure Type Red Polypropylene Cap
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'