missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethyleneimine, 50% Aqueous Solution, MP Biomedicals™
80.20 € - 370.00 €
Chemical Identifiers
CAS | 9002-98-6 |
---|---|
Molecular Formula | ((C2H5N)zC2H4N)y(C2H5N)x |
Molecular Weight (g/mol) | 43.07 |
MDL Number | MFCD00084427 |
InChI Key | NOWKCMXCCJGMRR-UHFFFAOYSA-N |
Synonym | ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin |
PubChem CID | 9033 |
ChEBI | CHEBI:30969 |
IUPAC Name | aziridine |
SMILES | *-CCNCCN(-*)CCN-* |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11495362
|
MP Biomedicals
0219544480 |
100 mL |
80.20 €
100mL |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||||
11405372
|
MP Biomedicals
0219544490 |
500 mL |
197.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11415372
|
MP Biomedicals
0219544491 |
1 L |
370.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
9002-98-6 | |
43.07 | |
NOWKCMXCCJGMRR-UHFFFAOYSA-N | |
9033 | |
aziridine |
((C2H5N)zC2H4N)y(C2H5N)x | |
MFCD00084427 | |
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin | |
CHEBI:30969 | |
*-CCNCCN(-*)CCN-* |
Specifications
9002-98-6 | |
((C2H5N)zC2H4N)y(C2H5N)x | |
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin | |
*-CCNCCN(-*)CCN-* | |
43.07 | |
CHEBI:30969 | |
Polyethyleneimine |
Viscous Liquid | |
MFCD00084427 | |
NOWKCMXCCJGMRR-UHFFFAOYSA-N | |
aziridine | |
9033 | |
50,000 to 100,000 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Polyethyleneimine, 50% aqueous solution
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
- H302-Harmful if swallowed.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : polyethyleneimine
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title