Learn More
Polyethyleneimine, branched, M.W. 10,000, 99%, Thermo Scientific Chemicals
73.77 € - 801.99 €
Chemical Identifiers
CAS | 9002-98-6 |
---|---|
Molecular Formula | ((C2H5N)zC2H4N)y(C2H5N)x |
Molecular Weight (g/mol) | 43.07 |
MDL Number | MFCD00084427 |
InChI Key | NOWKCMXCCJGMRR-UHFFFAOYSA-N |
Synonym | ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin |
PubChem CID | 9033 |
ChEBI | CHEBI:30969 |
IUPAC Name | aziridine |
SMILES | *-CCNCCN(-*)CCN-* |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11363898
|
Thermo Scientific Alfa Aesar
040331.14 |
25 g |
73.77 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11383898
|
Thermo Scientific Alfa Aesar
040331.22 |
100 g |
208.04 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11373898
|
Thermo Scientific Alfa Aesar
040331.36 |
500 g |
801.99 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Polyethyleneimine, branched, M.W. 10,000 acts as an auxiliary agent. It is used in paper-making, water treatment, detergents, adhesives and cosmetics. It is investigated as non-viral vector carriers in research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9002-98-6 | |
43.07 | |
NOWKCMXCCJGMRR-UHFFFAOYSA-N | |
9033 | |
aziridine |
((C2H5N)zC2H4N)y(C2H5N)x | |
MFCD00084427 | |
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin | |
CHEBI:30969 | |
*-CCNCCN(-*)CCN-* |
Specifications
Yellow | |
Liquid | |
MFCD00084427 | |
Soluble in toluene,methanol,ethanol,chloroform and xylene. Insoluble in water,alcohol,benzene,ethyl ether and acetone. | |
*-CCNCCN(-*)CCN-* | |
43.07 | |
CHEBI:30969 | |
Amine-like |
9002-98-6 | |
((C2H5N)zC2H4N)y(C2H5N)x | |
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin | |
NOWKCMXCCJGMRR-UHFFFAOYSA-N | |
aziridine | |
9033 | |
99% | |
Polyethyleneimine, branched, M.W. 10,000 |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'rtecsNumber' : TQ7915000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Polyethyleneimine, branched, M. W. 10,000
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Polyethyleneimine
RUO – Research Use Only