missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethyleneimine, branched, M.W. 50,000 - 100,000, 30% w/w aq. soln., Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Polyethyleneimine acts as a protein precipitant used to purify proteins. It is used as a chelating agent and as a scavenger for aldehydes and oxides. It is also used in detergents, paper industry, dyes, printing inks and in water treatment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Colorless to Yellow |
Physical Form | Liquid |
Solubility Information | Soluble in hot water at a low pH,methanol,ethanol,or chloroform. Insoluble in cold water,benzene,ethyl ether and acetone. |
Quantity | 100 g |
Concentration or Composition (by Analyte or Components) | 30% w/w aq. soln. |
Odor | Odorless |
Chemical Name or Material | Polyethyleneimine, Branched, M.W. 50,000 to 100,000 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'