missing translation for 'onlineSavingsMsg'
Learn More

Polyethyleneimine, ∽ M.N. 60,000, 50 wt.% aq. solution, branched, Thermo Scientific Chemicals

69.04 € - 380.90 €

Chemical Identifiers

CAS 9002-98-6, 7732-18-5
Molecular Formula ((C2H5N)zC2H4N)y(C2H5N)x
Molecular Weight (g/mol) 43.07
MDL Number MFCD00084427
InChI Key NOWKCMXCCJGMRR-UHFFFAOYSA-N
Synonym ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
PubChem CID 9033
ChEBI CHEBI:30969
IUPAC Name aziridine
SMILES *-CCNCCN(-*)CCN-*
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10720881
View Documents
Thermo Scientific Acros
178571000
100 g Glass bottle
69.04 €
100g
Please to purchase this item. Need a web account? Register with us today!
10499650
View Documents
Thermo Scientific Acros
178572500
250 g Glass bottle
133.67 €
250g
Please to purchase this item. Need a web account? Register with us today!
10592354
View Documents
Thermo Scientific Acros
178570010
1 kg Glass bottle
380.90 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9002-98-6, 7732-18-5
43.07
NOWKCMXCCJGMRR-UHFFFAOYSA-N
9033
aziridine
((C2H5N)zC2H4N)y(C2H5N)x
MFCD00084427
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
CHEBI:30969
*-CCNCCN(-*)CCN-*

Specifications

9002-98-6, 7732-18-5
100.0°C
(-CH2CH2NH-)x-(CH2CH2N(CH2CH2NH2)-)y
1.0700g/mL
Solubility in water: soluble.
*-CCNCCN(-*)CCN-*
43.07
CHEBI:30969
48 to 52% (solids)
1.07
Viscous Liquid
((C2H5N)zC2H4N)y(C2H5N)x
MFCD00084427
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
NOWKCMXCCJGMRR-UHFFFAOYSA-N
aziridine
9033
>110°C
Authentic
Polyethyleneimine, 50 wt. % Aqueous Solution, Average M.W. 50-60000
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : TQ7917000

missing translation for 'tsca' : TSCA

Product Identifier
  • Polyethyleneimine, 50 wt% aqueous solution, approx. M.N. 60,000

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.