missing translation for 'onlineSavingsMsg'
Learn More

Polyethyleneimine on silica beads, anion exchange resin, 20-40 mesh, Thermo Scientific Chemicals

For the separation of nucleotide mixtures

37.89 € - 382.00 €

Chemical Identifiers

CAS 9002-98-6
Molecular Formula ((C2H5N)zC2H4N)y(C2H5N)x
Molecular Weight (g/mol) 43.07
MDL Number MFCD00084427
InChI Key NOWKCMXCCJGMRR-UHFFFAOYSA-N
Synonym ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
PubChem CID 9033
ChEBI CHEBI:30969
IUPAC Name aziridine
SMILES *-CCNCCN(-*)CCN-*
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11349448
View Documents
Thermo Scientific Alfa Aesar
043302.AB
10 mL
37.89 €
10mL
In Stock
to see stock.
Add to basket
 
11339448
View Documents
Thermo Scientific Alfa Aesar
043302.AD
50 mL
108.00 €
50mL
In Stock
to see stock.
Add to basket
 
11329448
View Documents
Thermo Scientific Alfa Aesar
043302.AK
250 mL
382.00 €
250mL
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
Description

Description

Polyethyleneimine is used for protein and nucleic acid precipitation, transfection assays and to pretreat filters to increase the binding of proteins. It can also complex heavy metal ions. An effective permeabilizer of gram negative bacteria. Crosslinked polyethyleneimine is used as an enzyme carrier. It is used as a membrane lubricant, useful in filtering frozen cell membranes and for the separation of nucleotide mixtures. Polyethyleneimine cellulose in small columns will separate cyclic nucleotides from related compounds at the same electrolyte concentration that is used for thin layer chromatography on PEI cellulose-coated plates. It is also used in separation medium for thin layer chromatography, paper chromatography and Low pressure liquid chromatography. Polyethylenimine on silica gel is used for thin layer chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9002-98-6
43.07
NOWKCMXCCJGMRR-UHFFFAOYSA-N
9033
aziridine
((C2H5N)zC2H4N)y(C2H5N)x
MFCD00084427
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
CHEBI:30969
*-CCNCCN(-*)CCN-*

Specifications

9002-98-6
((C2H5N)zC2H4N)y(C2H5N)x
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
NOWKCMXCCJGMRR-UHFFFAOYSA-N
aziridine
9033
20 to 40 Mesh
Polyethyleneimine on silica beads anion exchange resin
Beads
MFCD00084427
Soluble in water.
*-CCNCCN(-*)CCN-*
43.07
CHEBI:30969
Hygroscopic
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.