missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethyleneimine on silica beads, anion exchange resin, benzylated, 40-200 mesh, Thermo Scientific™
Solid phase organic synthesis, affinity chromatography, catalyst support
21.00 € - 209.00 €
Chemical Identifiers
CAS | 9002-98-6 |
---|---|
Molecular Formula | ((C2H5N)zC2H4N)y(C2H5N)x |
Molecular Weight (g/mol) | 43.07 |
MDL Number | MFCD00084427 |
InChI Key | NOWKCMXCCJGMRR-UHFFFAOYSA-N |
Synonym | ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin |
PubChem CID | 9033 |
ChEBI | CHEBI:30969 |
IUPAC Name | aziridine |
SMILES | *-CCNCCN(-*)CCN-* |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
9002-98-6 | |
43.07 | |
NOWKCMXCCJGMRR-UHFFFAOYSA-N | |
9033 | |
aziridine |
((C2H5N)zC2H4N)y(C2H5N)x | |
MFCD00084427 | |
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin | |
CHEBI:30969 | |
*-CCNCCN(-*)CCN-* |
Specifications
9002-98-6 | |
((C2H5N)zC2H4N)y(C2H5N)x | |
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin | |
*-CCNCCN(-*)CCN-* | |
43.07 | |
CHEBI:30969 | |
Hygroscopic |
Powder | |
MFCD00084427 | |
NOWKCMXCCJGMRR-UHFFFAOYSA-N | |
aziridine | |
9033 | |
40 to 200 Mesh | |
Polyethyleneimine on silica beads anion exchange resin, Benzylated |
Safety and Handling
missing translation for 'tsca' : Yes
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title