Learn More
Macherey-Nagel™ Polygoprep™ 300 to 50 C18
Brand:  Macherey-Nagel™ 711730.1000
 View more versions of this product
Product Code. 10489151
  • 2845.00 € / 1000g
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
40 to 63 μm
POLYGOPREP 300 to 50
1 kg
Polygoprep
300 Å
2 to 8
POLYGOPREP C18
Liquid Chromatography