Learn More
Macherey-Nagel™ Polygoprep™ 60 to 12 C18
Brand:  Macherey-Nagel™ 711009.1000
 View more versions of this product
Product Code. 10002731
  • 1870.00 € / 1000g
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
10 to 15 μm
POLYGOPREP 60 to 18
2 to 8
POLYGOPREP C18
Polygoprep
Liquid Chromatography
60 Å
Liquid Chromatography
1 kg
350 m2/g
Silica