Learn More
Macherey-Nagel™ Polygoprep™ 60 to 80 C18
Brand:  Macherey-Nagel™ 711011.100
 View more versions of this product
Product Code. 10266261
  • 187.00 € / 100g
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
63 to 100 μm
POLYGOPREP 60 to 80
2 to 8
POLYGOPREP C18
Polygoprep
Liquid Chromatography
60 Å
Liquid Chromatography
0.1 kg
350 m2/g
Silica