Learn More
Macherey-Nagel™ Polygoprep™ 60 to 80 C18
Brand:  Macherey-Nagel™ 711011.1000
 View more versions of this product
Product Code. 10022741
  • 1375.00 € / 1000g
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
63 to 100 μm
POLYGOPREP 60 to 80
2 to 8
POLYGOPREP C18
Polygoprep
Liquid Chromatography
60 Å
Liquid Chromatography
1 kg
350 m2/g
Silica