Learn More
Macherey-Nagel™ Polygram CEL 300-10 UV254 Glass Plates
Brand:  Macherey-Nagel™ 808023
Product Code. 10154761
  • 195.00 € / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
25 Pk.
UV254
20 x 20 cm
Glass Plate
0.1 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Polygram CEL 300-10 UV254 Glass Plates > Plate Size: 200 x 200mm