missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ SIL G-25 / UV254 Glass Plates
Stable in almost all organic solvents. Macherey-Nagel™ SIL G-25 / UV254 Glass Plates are analytical plates with unmodified standard silica layers on glass for use in TLC. Feature a green fluorescence indicator for short-wave UV.
219.00 € - 320.00 €
Specifications
Material | Glass Plates with Unmodified Silica 60 Layers |
---|---|
Particle Size | 5 to 17 μm |
Pore Size | 60 Å |
Type | Glass TLC Plate |
Fluorescent Indicator | UV254 |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
12827951
|
Macherey-Nagel™
809027.200 |
5 x 10 cm |
320.00 €
Pack of 200 |
In Stock
Log in to see stock. |
||||||
12810951
|
Macherey-Nagel™
809028.100 |
2.5 x 7.5 cm |
219.00 €
Pack of 200 |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||||
Description
SIL G-25 / UV254 Glass Plates feature unmodified, standard-grade silica 60 layers with a green fluorescent indicator.
- Manganese-activated zinc silicate indicator with green fluorescence for short-wave UV (254nm)
- Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualization reagents
Specifications
Glass Plates with Unmodified Silica 60 Layers | |
60 Å | |
UV254 | |
∼500 m2/g |
5 to 17 μm | |
Glass TLC Plate | |
0.75 mL/g | |
0.25 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Macherey-Nagel™ SIL G-25 / UV254 Glass Plates