Learn More
Macherey-Nagel™ Standard SIL G Silica Layers on Polygram™ polyester sheets
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G silica layers on Polygram™ polyester sheets come in a variety of sizes. Select fluorescent indicator and choose from a variety of pack sizes.
41.05 € - 271.00 €
Specifications
Pore Size 60 Å
Type Polyester Sheet
Fluorescent Indicator UV254
Pore Volume 0.75 mL/g
Thickness (Metric) 0.2 mm
View More Specs
Products 6
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
10441193
View Documents
Macherey-Nagel™
805023
20 x 20 cm
151.00 €
Pack of 25
In Stock
to see stock.
Edge
Add to basket
10364501
View Documents
Macherey-Nagel™
805021
4 x 8 cm
41.05 €
Pack of 50
In Stock
to see stock.
Edge
Add to basket
10515751
View Documents
Macherey-Nagel™
805017
500 x 20 cm
144.00 €
1 roll
In Stock
to see stock.
Add to basket
10640493
View Documents
Macherey-Nagel™
805022
5 x 20 cm
82.10 €
Pack of 50
In Stock
to see stock.
Edge
Add to basket
12864491
View Documents
Macherey-Nagel™
805901
2.5 x 7.5 cm
152.00 €
Pack of 200
In Stock
to see stock.
Add to basket
10516721
View Documents
Macherey-Nagel™
805024
40 x 20 cm
271.00 €
Each
In Stock
to see stock.
Add to basket
Description
Show More
Specifications
Specifications
60 Å
UV254
0.2 mm
Polyester Sheet
0.75 mL/g
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Macherey-Nagel™ Standard SIL G Silica Layers on Polygram™ polyester sheets