missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygram SIL N-HR UV254 Polyester Sheets

Product Code. p-8012394 Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
5 x 20 cm
20 x 20 cm
missing translation for 'unitSize'
Pack of 25
Pack of 50
This item is not returnable. View return policy

Product Code. 10459751

missing translation for 'mfr': Macherey-Nagel™ 804023

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Pore Size 60 Å
Quantity 25 Pk.
Type Polyester Sheet
Fluorescent Indicator UV254
Pore Volume 0.75 mL/g
Thickness (Metric) 0.2 mm
Dimensions (L x W) 20 x 20 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.