missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Polymax 1040 (5° Tilt Angle)

The Polymax 1040 can be integrated into the incubation system, making temperature controlled applications possible.

Brand:  Heidolph™ 543-42205-06

Product Code. 17353030

  • 1885.00 € / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The 1000 series device can be integrated into the incubation system. This makes temperature-controlled applications with the Polymax 1040 possible.
  • With a load weight of up to 5 kg, all the required samples can be moved on this platform wave shaker. The speed can be adjusted between 2 to 50 rpm and the device can be operated either continuously or with a timer (max. 120 minutes).
Specifications

Specifications

230 V
UK Plug
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.