missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polymyxin B Sulfate MP Biomedicals
170.00 € - 824.00 €
Chemical Identifiers
CAS | 1405-20-5 |
---|---|
Molecular Formula | C48H84N16O17S |
Molecular Weight (g/mol) | 1189.36 |
MDL Number | MFCD00131991 |
InChI Key | HNDFYNOVSOOGDU-UHFFFAOYNA-N |
Synonym | Aerosporin |
PubChem CID | 133109994 |
IUPAC Name | N-[(2S)-4-amino-1-[[(2S,3R)-1-[[(2S)-4-amino-1-oxo-1-[[(3S,6S,18R)-6,9,18-tris(2-aminoethyl)-15-benzyl-3-[(1R)-1-hydroxyethyl]-12-(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptazacyclotricos-21-yl]amino]butan-2-yl]amino]-3-hydroxy-1-ox |
SMILES | OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11467660
|
MP Biomedicals
0210056505 |
5 mU |
170.00 €
5 micro units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11477660
|
MP Biomedicals
0210056510 |
10 mU |
251.00 €
10 micro units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11487660
|
MP Biomedicals
0210056525 |
25 mU |
491.00 €
25 micro units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11497660
|
MP Biomedicals
0210056550 |
50 mU |
824.00 €
50 micro units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
Description
Chemical Identifiers
1405-20-5 | |
1189.36 | |
HNDFYNOVSOOGDU-UHFFFAOYNA-N | |
133109994 | |
OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O |
C48H84N16O17S | |
MFCD00131991 | |
Aerosporin | |
N-[(2S)-4-amino-1-[[(2S,3R)-1-[[(2S)-4-amino-1-oxo-1-[[(3S,6S,18R)-6,9,18-tris(2-aminoethyl)-15-benzyl-3-[(1R)-1-hydroxyethyl]-12-(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptazacyclotricos-21-yl]amino]butan-2-yl]amino]-3-hydroxy-1-ox |
Specifications
Polymixin B Sulfate | |
1405-20-5 | |
MFCD00131991 | |
HNDFYNOVSOOGDU-UHFFFAOYNA-N | |
N-[(2S)-4-amino-1-[[(2S,3R)-1-[[(2S)-4-amino-1-oxo-1-[[(3S,6S,18R)-6,9,18-tris(2-aminoethyl)-15-benzyl-3-[(1R)-1-hydroxyethyl]-12-(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptazacyclotricos-21-yl]amino]butan-2-yl]amino]-3-hydroxy-1-ox | |
133109994 | |
White |
Powder | |
C48H84N16O17S | |
Aerosporin | |
OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O | |
1189.36 | |
1385.61 | |
Polymyxin B Sulfate, Aerosporin Activity: >6, 000 u/mg |
Safety and Handling
Recommended Storage : Store at 2° to 8°C.
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Polymyxin B Sulfate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : polymyxin B sulphate