missing translation for 'onlineSavingsMsg'
Learn More

Polyphosphoric Acid, 105% H3PO4 Basis, Honeywell Fluka™

Product Code. 15655380
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

∼105% H3PO4 basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8017-16-1
Molecular Formula (HO3P)n
MDL Number MFCD00084480
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name Polyphosphoric acid
SMILES [H]OP(O)(O)=O

Specifications

pH Acidic
Color Colorless
Melting Point 16°C
Boiling Point 300°C
Density 2.06g/cm3 at 20°C
Odor Odorless
Packaging Poly Bottle
Quantity 1 L
Linear Formula Hn+2PnO3n+1
Physical Form Liquid
UN Number UN3264
Percent Purity 105% (H3PO4 basis)
Chemical Name or Material Polyphosphoric acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.