missing translation for 'onlineSavingsMsg'
Learn More

Polyphosphoric acid, ca 84% (as phosphorus pentoxide), Thermo Scientific Chemicals

Product Code. 11373706
Click to view available options
Quantity:
100 mL
500 mL

Polyphosphoric acid is used as a dehydrating agent, cyclizing agent, acylating agent, analytical reagent and as a ring closing reagent. It is used in the production of phosphate esters, acid phosphates and pharmaceuticals. It is involved in metal treatment and as an asphalt and a bitumen additive. Furthermore, it is employed for the determination of tryptophan and used a reagent for the alkylation and acylation reactions of aromatic and alkenic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polyphosphoric acid is used as a dehydrating agent, cyclizing agent, acylating agent, analytical reagent and as a ring closing reagent. It is used in the production of phosphate esters, acid phosphates and pharmaceuticals. It is involved in metal treatment and as an asphalt and a bitumen additive. Furthermore, it is employed for the determination of tryptophan and used a reagent for the alkylation and acylation reactions of aromatic and alkenic compounds.

Solubility
Miscible with water.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with bases. It reacts with water violently to generate heat and phosphoric acid.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 8017-16-1
Molecular Formula (HO3P)n
MDL Number MFCD00084480
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name Polyphosphoric acid
SMILES [H]OP(O)(O)=O

Especificaciones

Density 2.1
Odor Odorless
Sensitivity Hygroscopic
Quantity 100 mL
UN Number UN3264
Merck Index 14,7580
Solubility Information Miscible with water.
Percent Purity ∼84%
Assay (as phosphorus pentoxide)
Chemical Name or Material Polyphosphoric acid
compliance-icons
Nombre del producto
  • Polyphosphoric acid
Palabra de advertencia
  • Danger
Clasificación
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Indicaciones de peligro
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Otros peligros
  • MIXTURE LIST-Contains : Polyphosphoric acid

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado