missing translation for 'onlineSavingsMsg'
Learn More

Polyphosphoric acid, ca 84% (as phosphorus pentoxide), Thermo Scientific Chemicals

Product Code. 11383706
Click to view available options
Quantity:
100 mL
500 mL

Polyphosphoric acid is used as a dehydrating agent, cyclizing agent, acylating agent, analytical reagent and as a ring closing reagent. It is used in the production of phosphate esters, acid phosphates and pharmaceuticals. It is involved in metal treatment and as an asphalt and a bitumen additive. Furthermore, it is employed for the determination of tryptophan and used a reagent for the alkylation and acylation reactions of aromatic and alkenic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polyphosphoric acid is used as a dehydrating agent, cyclizing agent, acylating agent, analytical reagent and as a ring closing reagent. It is used in the production of phosphate esters, acid phosphates and pharmaceuticals. It is involved in metal treatment and as an asphalt and a bitumen additive. Furthermore, it is employed for the determination of tryptophan and used a reagent for the alkylation and acylation reactions of aromatic and alkenic compounds.

Solubility
Miscible with water.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with bases. It reacts with water violently to generate heat and phosphoric acid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8017-16-1
Molecular Formula (HO3P)n
MDL Number MFCD00084480
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name Polyphosphoric acid
SMILES [H]OP(O)(O)=O

Specifications

Density 2.1
Odor Odorless
Sensitivity Hygroscopic
Quantity 500 mL
UN Number UN3264
Merck Index 14,7580
Solubility Information Miscible with water.
Percent Purity ∼84%
Assay (as phosphorus pentoxide)
Chemical Name or Material Polyphosphoric acid
compliance-icons
Product Identifier
  • Polyphosphoric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Superphosphoric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.