missing translation for 'onlineSavingsMsg'
Learn More

Polyphosphoric Acid, Reagent grade, 115% H3PO4 basis, Honeywell™

Product Code. 15631830
Click to view available options
Quantity:
25 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 115% H3PO4 basis

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8017-16-1
Molecular Formula (HO3P)n
MDL Number MFCD00084480
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name Polyphosphoric acid
SMILES [H]OP(O)(O)=O

Specifications

Color Colorless
Boiling Point 300°C
Density 2.06g/mL at 25°C
Packaging Poly Bottle
Quantity 25 g
Linear Formula Hn+2PnO3n+1
Physical Form Liquid
UN Number UN3264
Percent Purity 115% (H3PO4 basis)
Grade Reagent
Chemical Name or Material Polyphosphoric Acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.