Learn More
Fisherbrand™ Polypropylene 96 Well Microplate
Manufactured from inert polypropylene for thermal and solvent resistant qualities. Fisherbrand™ Polypropylene 96 Well Microplates are designed with a conical well base for superior sample concentration, reconstitution and centrifugation.
Brand:  Fisherbrand™ 219009
Encompass_Preferred
Product Code. 11511963
  • 273.05 € / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Unique, patented sealing cap allows penetration of a needle through the cap into each well, minimising coring of the needle as the base of each well of the cap has been reduced in thickness
  • Small radius on all corners to prevent sample precipitation and improve concentration
  • DNase and RNase free
  • Applications include SPE, HPLC, MS, liquid handling, automation, robotics
Specifications
Specifications
Standard
Polypropylene
2 mL
Yes
Yes
8 x 12
96
Square, Conical Bottom (V-well)
50
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Polypropylene 96 Well Microplate