Learn More
Hirschmann™ Polypropylene Thread Adapter
Polypropylene (PP) Threaded adapter for adaptation on the most common bottles.
Brand:  Hirschmann™ 9313505
 View more versions of this product
Product Code. 10380201
  • 15.35 € / Each
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 9313505/9313506: For use with ceramus™ -classic.
  • 9313527: For use with ceramus™, solarus™ and opus™.
Specifications
Specifications
Adapter
Polypropylene
A 32 to A 38