Learn More
Buerkle™ Polypropylene Barrel Screw Joint
Buerkle™ Polypropylene Barrel Screw Joint for secure connection in the barrel in three different thread models.
46.80 € - 66.25 €
Specifications
Material Polypropylene
Type Barrel Screw Joint
For Use With PTFE Barrel Pump
View More Specs
Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10309780
View Documents
Buerkle™
5600-3170
Mauser 2 in., Coarse Outer Thread
58.30 €
Each
In Stock
to see stock.
Add to basket
10625571
View Documents
Buerkle™
5600-3130
R 2 in., Fine Steel Outer Thread
46.80 €
Each
In Stock
to see stock.
Add to basket
10126370
View Documents
Buerkle™
5600-3180
Tri-Sure, Coarse Outer Thread
66.25 €
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Barrel screw joint made of PP for secure connection in the barrel in three different thread models
  • For drawing off liquids that are not flammable
  • The container connectors make decanting even easier and safer with their more secure connection to barrels
  • The connectors secure the pump in the barrel, enable a vertical position change and allow the required air exchange
Specifications
Specifications
Polypropylene
Barrel Screw Joint
PTFE Barrel Pump