missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Polypropylene Burette Clamps
To be mounted on support rods of 8 to 14mm dia. Easily affixes the burette by clamping with a non-corroding spring.
43.30 € - 47.15 €
Specifications
Color | White |
---|---|
Material | Polypropylene |
No. per Pack | 1 Each |
Type | Buret Holder |
Description
- Polypropylene construction
- White
- For use with support rods 8 to 14mm dia.
Specifications
White | |
Polypropylene | |
1 Each | |
Buret Holder |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title