Learn More
Euro Protection™ Polypropylene Bouffant Cap Product Code.: 12043266
In Stock
to see stock.
Add to basket
Euro Protection™ Polypropylene Bouffant Cap
Product Code. 12043266
White
Click to view available options
Color:
Blue
White
Product Code. 12043266
Brand: Euro Protection™ 45610
In Stock
to see stock.
Edge
Add to basket
Add to basket
Bouffant cap prevents stray hairs from contaminating the work space or laboratory where sterility is essential.
  • Round and elastic
  • Polypropylene
  • Blue or white
  • Bouffant style to cover hair
Specifications
Product Type Bouffant
Size One Size
Material Polypropylene
Color White
Disposable Single-use
Model 45610
Quantity 100/Pk.
Diameter (Metric) 53 cm
Product Content Correction
Product Title
Euro Protection™ Polypropylene Bouffant Cap >