missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Polypropylene Clear Disposal Bags
Made of polypropylene foil for sterilization in the steam autoclave. Greiner Bio-One™ is ideal for the hygienic disposal of contaminated laboratory items.
Specifications
Autoclavable | Autoclavable |
---|---|
Color | Clear |
Disposable | Single-use |
For Use With (Application) | Ideal for the hygienic disposal of contaminated laboratory items |
Material | Polypropylene |
Description
- Oxygen transport rate of 50 μm PP foil (+23°C): 2380 cm³/(m² x 24h x bar)
- Water vapour transport rate of 50 μm PP foil (+23°C, 85% air humidity): 1.54 g/(m² x 24h)
- Available on request with imprint "Biohazard" if a sufficient number is ordered
- Foil thickness: 0.05mm
- Autoclavable
- Steam autoclaves: Max 134°C, 2bar/30min
- Hot-air sterilizers: Max 140°C, 120min
- Temperature range: 0°C to 140°C
- For single use only
Specifications
Autoclavable | |
Single-use | |
Polypropylene |
Clear | |
Ideal for the hygienic disposal of contaminated laboratory items | |
0.050 mm |