Learn More
DWK Life Sciences Kimble Chase™ Polypropylene Closure with Ultrasonically Welded PTFE Liner
For use with screw-thread culture and centrifuge tubes
Brand:  DWK Life Sciences 73808-15415
 View more versions of this product
Product Code. 11532672
  • 631.00 € / Pack of 288
Estimated Shipment: 08-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Provides superior closure system capable of maintaining its liner through multiple autoclavable operations
  • PTFE-coated surface of exposed liner is chemical-resistant and compatible with sensitive, corrosive and volatile solvents
TRUSTED_SUSTAINABILITY
Specifications
Specifications
15-415
Screw Cap
Polypropylene
EPA Water Analysis Vials, Screw Thread Culture and Centrifuge Tubes
PTFE, Rubber
Autoclavable
For repeated autoclaving, to provide superior closure system