missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Polypropylene Conical Containers
Quality sampling tools for secure sample retrieval and shipment
Brand: Corning™ PC400-03
104.02 EUR valid until 2024-12-31
Use promo code "21693" to get your promotional price.
This item is not returnable.
View return policy
Description
- Lightweight and break-resistant
- Transparent or white Polypropylene (PP)
- Assembled or non-assembled
Specifications
Polypropylene | |
13.5 oz. | |
Natural | |
62 mm | |
185/Pk. |
Sterile | |
400 mL | |
Natural | |
185/Cs. |