Learn More
Kartell™ Polypropylene Conical Erlenmeyer Flasks
Translusent in design to allow one to view the mixture with ease. Kartell™ Polypropylene Conical Erlenmeyer Flasks are a simple flask that has a variety of applications in both chemistry and biology.
Brand:  Kartell™ 0146500
 View more versions of this product
Product Code. 10633092
  • 74.15 € / Pack of 4
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Translusent in design to allow one to view the mixture with ease. Kartell™ Polypropylene Conical Erlenmeyer Flasks are a simple flask that has a variety of applications in both chemistry and biology.
  • For general laboratory use
  • Provided with standardized neck
  • Autoclavable
  • Suitable for foodstuff
Specifications
Specifications
Translucent
Polypropylene
34/35 NS
Conical
Yes
2000 mL
100 mL
4
Autoclavable